Assay Details
| Gene Symbol: LOC100762031 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643266.1 | XP_007641456.1 | 12 - 13 | 1882 | 115 | |
| XM_007643267.1 | XP_007641457.1 | 12 - 13 | 1678 | 115 | ||
| XM_007643268.1 | XP_007641458.1 | 12 - 13 | 1652 | 115 | ||
| GenBank mRNA | JI889815.1 | - | 2 - 3 | 244 | 115 | |
| JP037950.1 | - | 2 - 3 | 244 | 115 | ||
More Information
Set Membership: |
|

LOC100762031
Genomic Map