Assay Details
| Gene Symbol: LOC100762031 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643266.1 | XP_007641456.1 | 3 - 4 | 551 | 59 | |
| XM_007643267.1 | XP_007641457.1 | 3 - 4 | 347 | 59 | ||
| XM_007643268.1 | XP_007641458.1 | 3 - 4 | 321 | 59 | ||
| GenBank mRNA | JI892757.1 | - | 3 - 4 | 535 | 59 | |
| JP053671.1 | - | 3 - 4 | 534 | 59 | ||
More Information
Set Membership: |
|

LOC100762031
Genomic Map