Assay Details
| Gene Symbol: Cnrip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502379.2 | XP_003502427.1 | 2 - 3 | 666 | 60 | |
| GenBank mRNA | JI888983.1 | - | 2 - 3 | 251 | 60 | |
| JP053821.1 | - | 2 - 3 | 434 | 60 | ||
More Information
Set Membership: |
|

Cnrip1
Genomic Map