Assay Details
| Gene Symbol: LOC100765644 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506111.2 | XP_003506159.2 | 3 - 4 | 1402 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100765644| Gene Symbol: LOC100765644 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506111.2 | XP_003506159.2 | 3 - 4 | 1402 | 102 | |
Set Membership: |
|