Assay Details
| Gene Symbol: Erbb2ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007639426.1 | XP_007637616.1 | 14 - 15 | 1335 | 87 | |
| XM_007639432.1 | XP_007637622.1 | 14 - 15 | 1335 | 87 | ||
| XM_007639434.1 | XP_007637624.1 | 14 - 15 | 1335 | 87 | ||
| XM_007639441.1 | XP_007637631.1 | 14 - 15 | 1335 | 87 | ||
| XM_007639446.1 | XP_007637636.1 | 14 - 15 | 1335 | 87 | ||
| XM_007639454.1 | XP_007637644.1 | 14 - 15 | 1335 | 87 | ||
| XM_007639463.1 | XP_007637653.1 | 14 - 15 | 1335 | 87 | ||
| GenBank mRNA | JI885126.1 | - | 15 - 16 | 1416 | 87 | |
| JP047696.1 | - | 15 - 16 | 1416 | 87 | ||
More Information
Set Membership: |
|

Erbb2ip
Genomic Map