Assay Details
| Gene Symbol: Sub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644232.1 | XP_007642422.1 | 1 - 2 | 163 | 84 | |
| GenBank mRNA | JI871443.1 | - | 2 - 3 | 123 | 84 | |
More Information
Set Membership: |
|

Sub1
Genomic Map