Assay Details
| Gene Symbol: Nt5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643369.1 | XP_007641559.1 | 1 - 2 | 224 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nt5c| Gene Symbol: Nt5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643369.1 | XP_007641559.1 | 1 - 2 | 224 | 57 | |
Set Membership: |
|