Assay Details
| Gene Symbol: Eltd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496430.2 | XP_003496478.1 | 6 - 7 | 472 | 80 | |
| XM_003496431.2 | XP_003496479.1 | 3 - 4 | 316 | 80 | ||
More Information
Set Membership: |
|

Eltd1
Genomic Map