Assay Details
Gene Symbol: LOC100751280 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504553.2 | XP_003504601.1 | 10 - 11 | 1541 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
