Assay Details
| Gene Symbol: Pter | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505037.2 | XP_003505085.1 | 4 - 5 | 1045 | 75 | |
| XM_007645542.1 | XP_007643732.1 | 3 - 4 | 778 | 75 | ||
| XM_007645543.1 | XP_007643733.1 | 5 - 6 | 1130 | 75 | ||
| XM_007645544.1 | XP_007643734.1 | 4 - 5 | 880 | 75 | ||
| XM_007645545.1 | XP_007643735.1 | 3 - 4 | 778 | 75 | ||
| XM_007645546.1 | XP_007643736.1 | 3 - 4 | 778 | 75 | ||
| XM_007645547.1 | XP_007643737.1 | 3 - 4 | 778 | 75 | ||
| GenBank mRNA | JI878796.1 | - | 2 - 3 | 722 | 75 | |
More Information
Set Membership: |
|

Pter
Genomic Map