Assay Details
| Gene Symbol: Adh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007607669.1 | XP_007605859.1 | 5 - 6 | 658 | 58 | |
| GenBank mRNA | JI892485.1 | - | 5 - 6 | 625 | 58 | |
More Information
Set Membership: |
|

Adh5
Genomic Map