Assay Details
| Gene Symbol: Emr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642457.1 | XP_007640647.1 | 13 - 14 | 1700 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Emr1| Gene Symbol: Emr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642457.1 | XP_007640647.1 | 13 - 14 | 1700 | 63 | |
Set Membership: |
|