Assay Details
| Gene Symbol: Myoz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496718.1 | XP_003496766.1 | 4 - 5 | 659 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myoz1| Gene Symbol: Myoz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496718.1 | XP_003496766.1 | 4 - 5 | 659 | 61 | |
Set Membership: |
|