Assay Details
| Gene Symbol: Gtf2a1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499540.1 | XP_003499588.1 | 3 - 4 | 241 | 62 | |
| XM_003499541.1 | XP_003499589.1 | 2 - 3 | 139 | 62 | ||
More Information
Set Membership: |
|

Gtf2a1l
Genomic Map