Assay Details
| Gene Symbol: Rab7l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501207.2 | XP_003501255.1 | 2 - 3 | 606 | 86 | |
| GenBank mRNA | JP056939.1 | - | 2 - 3 | 345 | 86 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab7l1| Gene Symbol: Rab7l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501207.2 | XP_003501255.1 | 2 - 3 | 606 | 86 | |
| GenBank mRNA | JP056939.1 | - | 2 - 3 | 345 | 86 | |