Assay Details
| Gene Symbol: Cactin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646986.1 | XP_007645176.1 | 5 - 6 | 1136 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cactin| Gene Symbol: Cactin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646986.1 | XP_007645176.1 | 5 - 6 | 1136 | 69 | |
Set Membership: |
|