Assay Details
| Gene Symbol: F2rl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498712.2 | XP_003498760.1 | 1 - 2 | 200 | 68 | |
| XM_007632111.1 | XP_007630301.1 | 1 - 2 | 200 | 68 | ||
More Information
Set Membership: |
|

F2rl2
Genomic Map