Assay Details
Gene Symbol: Mocs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502231.2 | XP_003502279.1 | 5 - 6 | 636 | 93 | |
GenBank mRNA | JI875579.1 | - | 5 - 6 | 632 | 93 | |
JP055919.1 | - | 5 - 6 | 631 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
