Assay Details
| Gene Symbol: Ccdc129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503917.1 | XP_003503965.1 | 7 - 8 | 1172 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc129| Gene Symbol: Ccdc129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503917.1 | XP_003503965.1 | 7 - 8 | 1172 | 103 | |
Set Membership: |
|