Assay Details
Gene Symbol: Rlbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512021.1 | XP_003512069.1 | 1 - 2 | 145 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
