Assay Details
| Gene Symbol: Dnajc19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647287.1 | XP_007645477.1 | 6 - 7 | 571 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnajc19| Gene Symbol: Dnajc19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647287.1 | XP_007645477.1 | 6 - 7 | 571 | 66 | |
Set Membership: |
|