Assay Details
| Gene Symbol: LOC100771548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653528.1 | XP_007651718.1 | 8 - 9 | 2151 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100771548| Gene Symbol: LOC100771548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653528.1 | XP_007651718.1 | 8 - 9 | 2151 | 69 | |
Set Membership: |
|