Assay Details
| Gene Symbol: LOC100768534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645456.1 | XP_007643646.1 | 4 - 5 | 1987 | 70 | |
| XM_007645458.1 | XP_007643648.1 | 4 - 5 | 2010 | 70 | ||
| GenBank mRNA | JI895711.1 | - | 1 - 2 | 339 | 70 | |
| JP050643.1 | - | 1 - 2 | 339 | 70 | ||
More Information
Set Membership: |
|

LOC100768534
Genomic Map