Assay Details
| Gene Symbol: Slc39a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646387.1 | XP_007644577.1 | 3 - 4 | 413 | 61 | |
| GenBank mRNA | JI879247.1 | - | 3 - 4 | 254 | 61 | |
More Information
Set Membership: |
|

Slc39a7
Genomic Map