Assay Details
| Gene Symbol: Fam8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645998.1 | XP_007644188.1 | 3 - 4 | 408 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam8a1| Gene Symbol: Fam8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645998.1 | XP_007644188.1 | 3 - 4 | 408 | 108 | |
Set Membership: |
|