Assay Details
| Gene Symbol: Tspan9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496022.2 | XP_003496070.2 | 2 - 3 | 1103 | 61 | |
| XM_007646329.1 | XP_007644519.1 | 3 - 4 | 376 | 61 | ||
| XM_007646334.1 | XP_007644524.1 | 3 - 4 | 414 | 61 | ||
| XM_007646338.1 | XP_007644528.1 | 3 - 4 | 289 | 61 | ||
More Information
Set Membership: |
|

Tspan9
Genomic Map