Assay Details
| Gene Symbol: Ebi3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501740.2 | XP_003501788.1 | 4 - 5 | 602 | 75 | |
| XM_007642403.1 | XP_007640593.1 | 4 - 5 | 614 | 75 | ||
More Information
Set Membership: |
|

Ebi3
Genomic Map