Assay Details
| Gene Symbol: Pms1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513713.2 | XP_003513761.2 | 2 - 3 | 439 | 68 | |
| GenBank mRNA | JP050998.1 | - | 2 - 3 | 439 | 68 | |
More Information
Set Membership: |
|

Pms1
Genomic Map