Assay Details
| Gene Symbol: Tert | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515019.2 | XP_003515067.2 | 4 - 5 | 2644 | 61 | |
| XM_007655042.1 | XP_007653232.1 | 5 - 6 | 2644 | 61 | ||
| XM_007655043.1 | XP_007653233.1 | 5 - 6 | 2644 | 61 | ||
More Information
Set Membership: |
|

Tert
Genomic Map