Assay Details
| Gene Symbol: Taf5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653474.1 | XP_007651664.1 | 2 - 3 | 174 | 67 | |
| XM_007653475.1 | XP_007651665.1 | 1 - 2 | 155 | 67 | ||
| GenBank mRNA | JI884345.1 | - | 1 | 57 | 67 | |
| JP051380.1 | - | 1 | 57 | 67 | ||
More Information
Set Membership: |
|

Taf5l
Genomic Map