Assay Details
| Gene Symbol: Prpf18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496413.2 | XP_003496461.1 | 1 - 2 | 206 | 68 | |
| GenBank mRNA | JI885696.1 | - | 1 - 2 | 208 | 68 | |
| JP049593.1 | - | 1 - 2 | 208 | 68 | ||
More Information
Set Membership: |
|

Prpf18
Genomic Map