Assay Details
| Gene Symbol: Mmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498186.2 | XP_003498234.1 | 2 - 3 | 133 | 63 | |
| XM_007624037.1 | XP_007622227.1 | 2 - 3 | 133 | 63 | ||
| XM_007624049.1 | XP_007622239.1 | 2 - 3 | 133 | 63 | ||
More Information
Set Membership: |
|

Mmd
Genomic Map