Assay Details
| Gene Symbol: Ace3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502984.1 | XP_003503032.1 | 5 - 6 | 628 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ace3| Gene Symbol: Ace3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502984.1 | XP_003503032.1 | 5 - 6 | 628 | 94 | |
Set Membership: |
|