Assay Details
| Gene Symbol: Panx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506484.2 | XP_003506532.1 | 2 - 3 | 677 | 61 | |
| GenBank mRNA | JI883140.1 | - | 2 - 3 | 298 | 61 | |
| JP060882.1 | - | 2 - 3 | 298 | 61 | ||
More Information
Set Membership: |
|

Panx1
Genomic Map