Assay Details
| Gene Symbol: LOC100751755 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506888.2 | XP_003506936.1 | 5 - 6 | 731 | 58 | |
| GenBank mRNA | JI868390.1 | - | 2 - 3 | 189 | 58 | |
| JP039759.1 | - | 2 - 3 | 189 | 58 | ||
More Information
Set Membership: |
|

LOC100751755
Genomic Map