Assay Details
| Gene Symbol: Stx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510982.2 | XP_003511030.1 | 2 - 3 | 265 | 63 | |
| GenBank mRNA | JI874817.1 | - | 2 - 3 | 377 | 63 | |
| JP062548.1 | - | 2 - 3 | 377 | 63 | ||
More Information
Set Membership: |
|

Stx4
Genomic Map