Assay Details
| Gene Symbol: Plac8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511833.2 | XP_003511881.1 | 4 - 5 | 545 | 70 | |
| GenBank mRNA | JI883317.1 | - | 4 - 5 | 458 | 70 | |
| JP055297.1 | - | 4 - 5 | 464 | 70 | ||
More Information
Set Membership: |
|

Plac8
Genomic Map