Assay Details
| Gene Symbol: Ube2j1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641094.1 | XP_007639284.1 | 6 - 7 | 660 | 66 | |
| GenBank mRNA | JI863793.1 | - | 6 - 7 | 662 | 66 | |
More Information
Set Membership: |
|

Ube2j1
Genomic Map