Assay Details
| Gene Symbol: LOC100771249 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495663.1 | XP_003495711.1 | 5 - 6 | 1173 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100771249| Gene Symbol: LOC100771249 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495663.1 | XP_003495711.1 | 5 - 6 | 1173 | 64 | |
Set Membership: |
|