Assay Details
| Gene Symbol: Tfpi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496969.2 | XP_003497017.2 | 4 - 5 | 765 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tfpi2| Gene Symbol: Tfpi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496969.2 | XP_003497017.2 | 4 - 5 | 765 | 70 | |
Set Membership: |
|