Assay Details
| Gene Symbol: Hsh2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499085.2 | XP_003499133.1 | 4 - 5 | 846 | 76 | |
| XM_007638238.1 | XP_007636428.1 | 5 - 6 | 813 | 76 | ||
| XM_007638245.1 | XP_007636435.1 | 5 - 6 | 743 | 76 | ||
| XM_007638250.1 | XP_007636440.1 | 5 - 6 | 728 | 76 | ||
More Information
Set Membership: |
|

Hsh2d
Genomic Map