Assay Details
| Gene Symbol: Fabp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510074.2 | XP_003510122.1 | 1 - 2 | 172 | 74 | |
| GenBank mRNA | JI884147.1 | - | 1 - 2 | 309 | 74 | |
| JP054072.1 | - | 1 - 2 | 308 | 74 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fabp4| Gene Symbol: Fabp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510074.2 | XP_003510122.1 | 1 - 2 | 172 | 74 | |
| GenBank mRNA | JI884147.1 | - | 1 - 2 | 309 | 74 | |
| JP054072.1 | - | 1 - 2 | 308 | 74 | ||