Assay Details
Gene Symbol: Fcgr2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647410.1 | XP_007645600.1 | 2 - 3 | 402 | 65 | |
XM_007647411.1 | XP_007645601.1 | 3 - 4 | 535 | 65 | ||
GenBank mRNA | JP061796.1 | - | 2 - 3 | 248 | 65 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
