Assay Details
Gene Symbol: Fcgr2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647410.1 | XP_007645600.1 | 3 - 4 | 427 | 89 | |
XM_007647411.1 | XP_007645601.1 | 4 - 5 | 560 | 89 | ||
GenBank mRNA | JP061796.1 | - | 3 - 4 | 273 | 89 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
