Assay Details
Gene Symbol: Fcgr2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647410.1 | XP_007645600.1 | 5 - 6 | 942 | 75 | |
XM_007647411.1 | XP_007645601.1 | 6 - 7 | 1075 | 75 | ||
XM_007647412.1 | XP_007645602.1 | 4 - 5 | 728 | 75 | ||
GenBank mRNA | JP061796.1 | - | 5 - 6 | 788 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
