Assay Details
| Gene Symbol: Tcf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495072.2 | XP_003495120.1 | 12 - 13 | 1498 | 64 | |
| XM_007651019.1 | XP_007649209.1 | 12 - 13 | 1497 | 64 | ||
| GenBank mRNA | JI890674.1 | - | 12 - 13 | 1436 | 64 | |
| JP058979.1 | - | 12 - 13 | 1436 | 64 | ||
More Information
Set Membership: |
|

Tcf25
Genomic Map