Assay Details
| Gene Symbol: Pxylp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495485.2 | XP_003495533.1 | 4 - 5 | 557 | 82 | |
| GenBank mRNA | JI888537.1 | - | 4 - 5 | 618 | 82 | |
| JP047945.1 | - | 4 - 5 | 618 | 82 | ||
More Information
Set Membership: |
|

Pxylp1
Genomic Map