Assay Details
Gene Symbol: Tnfaip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496545.2 | XP_003496593.2 | 1 - 2 | 514 | 62 | |
XM_007651164.1 | XP_007649354.1 | 2 - 3 | 579 | 62 | ||
XM_007651169.1 | XP_007649359.1 | 2 - 3 | 580 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
