Assay Details
| Gene Symbol: Mfap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498030.2 | XP_003498078.1 | 4 - 5 | 775 | 70 | |
| GenBank mRNA | JI862941.1 | - | 4 - 5 | 776 | 70 | |
| JP041555.1 | - | 4 - 5 | 776 | 70 | ||
More Information
Set Membership: |
|

Mfap1
Genomic Map