Assay Details
| Gene Symbol: Sf3b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498632.2 | XP_003498680.1 | 1 - 2 | 85 | 57 | |
| GenBank mRNA | JI878801.1 | - | 1 - 2 | 93 | 57 | |
| JP054115.1 | - | 1 - 2 | 93 | 57 | ||
More Information
Set Membership: |
|

Sf3b4
Genomic Map